Learn More
Helios Italquartz 1234 DIN Standard Covers with Ring Handle, Transparent
1234 DIN standard covers with ring handle constructed of transparent, durable, resistant, and chemically inert quartz glass.
Marke:  Helios Italquartz 64P00605
Additional Details : Gewicht : 0.20000kg
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15828094
Menge Preis
1 91.67 CHF / 1 Stück
10 85.36 CHF / 1 Stück
Verfügbar ab
24-06-2021
Zum Warenkorb hinzufügen
Beschreibung
Beschreibung
  • Transparent
  • Include ring handle
  • Conform to 1234 DIN standard
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Spezifikation
Spezifikation
Transparent
CR 45-54-65 Crucible
Quartz
1234 DIN Standard Covers with Ring Handle