Learn More
Sartorius™ 11406 Serie, Sterile Membranfilter aus Cellulose Nitrat (CN) mit Gitternetz
11406 Series Gridded Sterile Cellulose Nitrate (CN) Membrane Filters have a very uniform pore structure and high non-specific adsorption. Single-packed or dispenser-ready, these individually wrapped filters are ideal for colony counting, particle testing, microscopy, and sterility testing.
Marke: Sartorius™ 11406--47----SCM
Additional Details : CAS : 7727-37-9 Gewicht : 0.01000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II
Beschreibung
- Product Type: Membrane Filter
- Material: Cellulose Nitrate
- Membrane Color/Grid Color: White/Black
- Pore Size: 0.45 μm
- Sterility: Sterile
- For Use With (Application): Bioburden/Microbial Enumeration, Food and Beverage, Environmental/Water Testing
Spezifikation
Weiß | |
SMART Dispenser-Membranfilter | |
Cellulosenitrat (CN) | |
0.45 μm | |
Zur Partikelsammlung | |
300/Pckg. | |
5 mil | |
SMART Membranfilter aus Cellulosenitrat | |
47 mm |
Single-use | |
Disc | |
Cellulosenitrat | |
Sterile | |
Schwarz | |
300 Pk. | |
130 μ m | |
1.9 in. | |
47 mm |