Learn More
Sartorius™ Nicht-sterile Membranfilter aus Cellulosenitrat (CN), Serie 11303
11303 Series Non-Sterile Cellulose Nitrate (CN) Membrane Filters are hydrophilic, have high flow rates thanks to their symmetrical structure and are compatible with aqueous solutions (pH 4 – 8), hydrocarbons, and several other organic solvents.
Marke: Sartorius™ 11303--13------N
Additional Details : Gewicht : 0.10000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II
Beschreibung
- High, non-specific absorption
- Hydrophilic, with high flow rates
- Available in a range of pore sizes and diameters
- Ideal for several laboratory applications including particle retention, cell capture, cell retention, and cell filtration
Spezifikation
Autoklavierbar | |
Weiß | |
Membranfilter | |
Cellulosenitrat | |
1,2 μm | |
200 ml/min/cm2/bar | |
100/Packung | |
5 mil | |
Membranfilter | |
13 mm |
Gehäuse 1bar | |
Single-use | |
Disc | |
Cellulosenitrat | |
Unsteril | |
Für viele allgemeine Laboranwendungen, bei denen eine Membran mit einer hohen unspezifischen Adsorption geeignet ist | |
100 Pk. | |
130 μm | |
0.5 in. | |
13 mm |