Learn More
1-Methyl-1,2,3,4-Tetrahydroisochinolin, 95 %, Thermo Scientific Chemicals
Beschreibung
1-Methyl-1,2,3,4-tetrahydroisoquinoline (1MeTIQ) is an endogenous antidepressant and parkinsonism-preventing substance that demonstrates neuroprotectiveactivity. Following systemic administration in rats, 1-Methyl-1,2,3,4-tetrahydroisoquinoline produces antidepressant-like effect similar to the effect of imipramine. 1MeTIQ is a reversible short-acting moderate inhibitor of MAO A/B. Also known as 1,2,3,4-Tetrahydro-1-methylisoquinoline, 1MeTIQ. It is an important raw material and intermediate for revaprazan and pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Schwer in Wasser löslich.
Hinweise
Im Tiefkühlgerät bei (-20 °C) lagern. Behälter dicht verschlossen lassen. Von Oxidationsmitteln entfernt aufbewahren.
Spezifikation
Chemischer Name oder Material | 1-Methyl-1,2,3,4-tetrahydroisoquinoline |
Farbe | Gelb |
Siedepunkt | 119°C (18 mmHg) |
Summenformel | C10H13N |
Menge | 1 g |
Beilstein | 82140 |
Synonym | 1-m-thiq, 1-methyl-1,2,3,4-tetrahydro-isoquinoline, 1,2,3,4-tetrahydro-1-methylisoquinoline, chembl95456, isoquinoline, tetrahydromethyl, n-methyl,tetrahydro-isoquinoline, isoquinoline, 1,2,3,4-tetrahydro-1-methyl, 1metiq, pubchem13018, acmc-1aktb |
Löslichkeitsinformationen | Slightly soluble in water. |
InChI-Schlüssel | QPILYVQSKNWRDD-UHFFFAOYNA-N |
SMILES | CC1C2=CC=CC=C2CCN1 |
Mehr anzeigen |
RUO – Research Use Only