Learn More
1-Acenaphthenon, TRC
Specifications
Specifications
| Chemischer Name oder Material | 1-Acenaphthenone |
| CAS | 2235-15-6 |
| Summenformel | C12 H8 O |
| Menge | 1 g |
| Synonym | 1-Acenaphthone, 2-Acenaphthone, Acenaphthenone, Acenaphthylen-1-one, NSC 18756, NSC 22341, 1-Acenaphthenone (6CI,7CI,8CI), 1,2-Dihydroacenaphthylen-1-one, 2H-Acenaphthylen-1-one |
| InChI-Formel | InChI=1S/C12H8O/c13-11-7-9-5-1-3-8-4-2-6-10(11)12(8)9/h1-6H,7H2 |
| SMILES | O=C1Cc2cccc3cccc1c23 |
| IUPAC-Name | 2H-Acenaphthylen-1-on |
| Molekulargewicht (g/mol) | 168.19 |
| Formelmasse | 168.0575 |