Learn More
1-(3-Hydroxyphenyl)-2-(methylamino)ethanonhydrochlorid (Phenylephronhydrochlorid), Mikromol™
Beschreibung
1-(3-Hydroxyphenyl)-2-(methylamino)ethanonhydrochlorid (Phenylephronhydrochlorid)
Spezifikation
Spezifikation
| Chemischer Name oder Material | Phenylephrone Hydrochloride |
| CAS | 94240-17-2 |
| Bezeichnung der Analyten oder Bestandteile | Phenylephrone Hydrochloride |
| Versandbedingung | Room Temperature |
| Summenformel | C9 H11 N O2 . Cl H |
| Zertifizierung/Konformität | ISO 17025 |
| Synonym | Phenylephrine Hydrochloride Imp. C (EP), Phenylephrine Imp. C (EP), Phenylephrine USP Related Compound C, Phenylephrine USP RC C, 1-(3-Hydroxyphenyl)-2-(methylamino)ethanone Hydrochloride, Phenylephrine Hydrochloride Imp. C (EP) as Hydrochloride, Phenylephrone Hydrochloride, Ethanone, 1-(3-hydroxyphenyl)-2-(methylamino)-, hydrochloride (1:1), Ethanone, 1-(3-hydroxyphenyl)-2-(methylamino)-, hydrochloride (9CI), Phenylephrine Related Compound C as Hydrochloride, Phenylephrine Hydrochloride Impurity C as Hydrochloride, Phenylephrine Impurity C as Hydrochloride, Phenylephrine Impurity C as Hydrochloride |
| InChI-Formel | InChI=1S/C9H11NO2.ClH/c1-10-6-9(12)7-3-2-4-8(11)5-7;/h2-5,10-11H,6H2,1H3;1H |
| SMILES | Cl.CNCC(=O)c1cccc(O)c1 |
| IUPAC-Name | 1-(3-Hydroxyphenyl)-2-(methylamino)ethanon;hydrochlorid |
| Mehr anzeigen |