Learn More
1-(3-Ethyl-5,5,8,8-tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethanone, TRC
Artikelnummer. 30411720
Change view
Click to view available options
Menge:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
30411720 25 mg
30417668 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30411720 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCT29540525MG
om dit product te kopen Registreer vandaag om een webaccount aan te maken
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 88-29-9
Summenformel C18 H26 O
Molekulargewicht (g/mol) 258.4
Synonym 2'-Acetonaphthone, 3'-ethyl-5',6',7',8'-tetrahydro-5',5',8',8'-tetramethyl- (7CI,8CI), Versalide (6CI), 1-(3-Ethyl-5,6,7,8-tetrahydro-5,5,8,8-tetramethyl-2-naphthalenyl)ethanone, 1,1,4,4-Tetramethyl-6-ethyl-7-acetyl-1,2,3,4-tetrahydronaphthalene, 1,1,4,4-Tetramethyl-6-ethyl-7-acetyltetralin, 3'-Ethyl-5',6',7',8'-tetrahydro-5',5',8',8'-tetramethyl-2'-acetonaphthone, 7-Acetyl-6-ethyl-1,1,4,4-tetramethyltetralin, 7-Acetyl-6-ethyl-1,2,3,4-tetrahydro-1,1,4,4-tetramethylnaphthalene, AETT, Musk 36A, NSC 15342
IUPAC-Name 1-(3-ethyl-5,5,8,8-tetramethyl-6,7-dihydronaphthalen-2-yl)ethanone
SMILES CCc1cc2c(cc1C(=O)C)C(C)(C)CCC2(C)C
Spezifikation
Chemischer Name oder Material Versalide
Menge 25 mg
InChI-Formel InChI=1S/C18H26O/c1-7-13-10-15-16(11-14(13)12(2)19)18(5,6)9-8-17(15,3)4/h10-11H,7-9H2,1-6H3
Formelmasse 258.1984