missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethyladamantane, TRC

Artikelnummer. 30455209
Change view
Click to view available options
Menge:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge
30455209 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30455209 Supplier Toronto Research Chemicals Supplier No. TRCT795780250MG

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 707-35-7
Summenformel C13 H22
Molekulargewicht (g/mol) 178.31
Synonym Tricyclo[3.3.1.13,7]decane, 1,3,5-trimethyl-, Adamantane, 1,3,5-trimethyl- (7CI,8CI), 1,3,5-Trimethyltricyclo[3.3.1.13,7]decane, 1,3,5-Trimethyladamantane
IUPAC-Name 1,3,5-trimethyladamantane
SMILES CC12CC3CC(C)(CC(C)(C3)C1)C2

Specifications

Chemischer Name oder Material 1,3,5-Trimethyladamantane
Menge 250 mg
InChI-Formel InChI=1S/C13H22/c1-11-4-10-5-12(2,7-11)9-13(3,6-10)8-11/h10H,4-9H2,1-3H3
Formelmasse 178.1722
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.