Learn More
1,2-Bis(4-(benzo[d]oxazol-2-yl)phenyl)ethen, TRC
Specifications
Specifications
| Chemischer Name oder Material | 4,4'-Bis(2-benzoxazolyl)stilbene |
| CAS | 1533-45-5 |
| Summenformel | C28 H18 N2 O2 |
| Menge | 250 mg |
| Synonym | Benzoxazole, 2,2'-(vinylenedi-p-phenylene)bis- (7CI,8CI), 2,2'-(1,2-Ethenediyldi-4,1-phenylene)bis[benzoxazole], 2,2'-(Vinylenedi-p-phenylene)bisbenzoxazole, 4,4'-Bis(2-benzoxazolyl)stilbene, 4,4'-Di(benzoxazol-2-yl)stilbene, Benetex OB 1, C.I. 40674, C.I. Fluorescent Brightener 393, Eastobrite OB 1, Eastobrite OB 3, Fluorescent Brightener 393, OB 1, Optical Brightener 1, TM, TM (UV stabilizer), Uvitex OB 1, Uvitex OB-ONE, p,p'-Bis(2-benzoxazolyl)stilbene |
| InChI-Formel | InChI=1S/C28H18N2O2/c1-3-7-25-23(5-1)29-27(31-25)21-15-11-19(12-16-21)9-10-20-13-17-22(18-14-20)28-30-24-6-2-4-8-26(24)32-28/h1-18H |
| SMILES | o1c(nc2ccccc12)c3ccc(C=Cc4ccc(cc4)c5oc6ccccc6n5)cc3 |
| IUPAC-Name | 2-[4-[2-[4-(1,3-Benzoxazol-2-yl)phenyl]ethenyl]phenyl]-1,3-Benzoxazol |
| Molekulargewicht (g/mol) | 414.45 |
| Formelmasse | 414.1368 |