missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzisothiazol-3-on, 97 %, Thermo Scientific Chemicals

Catalog Number p-7040665
Click to view available options
Menge:
5 g
25 g
2634-33-5
C7H5NOS
151.183
MFCD00127753
DMSMPAJRVJJAGA-UHFFFAOYSA-N
1,2-benzisothiazol-3 2h-one, 1,2-benzisothiazolin-3-one, benzisothiazolone, proxel, benzo d isothiazol-3 2h-one, 1,2-benzisothiazoline-3-one, benzisothiazolinone, proxel pl, benzo d isothiazol-3-one, 2,3-dihydro-1,2-benzothiazol-3-one
17520
CHEBI:167099
1,2-Benzothiazol-3-on
C1=CC=C2C(=C1)C(=O)NS2
2634-33-5
C7H5NOS
151.183
MFCD00127753
DMSMPAJRVJJAGA-UHFFFAOYSA-N
1,2-benzisothiazol-3 2h-one, 1,2-benzisothiazolin-3-one, benzisothiazolone, proxel, benzo d isothiazol-3 2h-one, 1,2-benzisothiazoline-3-one, benzisothiazolinone, proxel pl, benzo d isothiazol-3-one, 2,3-dihydro-1,2-benzothiazol-3-one
17520
CHEBI:167099
1,2-Benzothiazol-3-on
C1=CC=C2C(=C1)C(=O)NS2

It is an antimicrobial agent and a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Anwendungen
Es handelt sich um ein antimikrobielles Mittel und ein pharmazeutisches Zwischenprodukt.

Löslichkeit
Löslich in Dichlormethan, Dimethylsulfoxid, Methanol.

Hinweise
Bei Raumtemperatur lagern. Von Hitze und Oxidationsmitteln fernhalten.
TRUSTED_SUSTAINABILITY
Chemischer Name oder Material 1,2-Benzisothiazol-3-one
Schmelzpunkt 154°C to 158°C
Menge 5 g
UN-Nummer UN3077
Löslichkeitsinformationen Soluble in dichloromethane,dimethyl sulfoxide,methanol.
Formelmasse 151.19
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'