CAS RN 7693-41-6
CAS RN 7693-41-6
IUPAC Name:
4-methoxyphenyl carbonochloridate
Synonyme:
4-methoxyphenyl carbonochloridate
Molekulargewicht (g/mol):
186.59
Summenformel:
C8H7ClO3
InChi Key:
CCFSGQKTSBIIHG-UHFFFAOYSA-N
SMILES:
COC1=CC=C(OC(Cl)=O)C=C1
1
–
1
von
1
Ergebnisse
1
4-Methoxyphenylchloroformat, 98 %, Thermo Scientific Chemicals
CAS: 7693-41-6 Summenformel: C8H7ClO3 Molekulargewicht (g/mol): 186.591 MDL-Nummer: MFCD00013258 InChI-Schlüssel: CCFSGQKTSBIIHG-UHFFFAOYSA-N Synonym: 4-methoxyphenyl chloroformate,carbonochloridic acid, 4-methoxyphenyl ester,4-methoxyphenylchloroformate,chloroformic acid 4-methoxyphenyl ester,acmc-1bfyr,p-methoxyphenyl chloroformate,4-methoxy-phenyl chloroformate,4-methoxy phenyl chloroformate,4-methoxyphenyl carbonochloridate PubChem CID: 82128 IUPAC-Name: (4-methoxyphenyl) carbonochloridat SMILES: COC1=CC=C(C=C1)OC(=O)Cl