CAS RN 7429-91-6
Dysprosiumlösung für ICP/MS 10 mg/L HNO₃- Matrix, Fisher chemical™
CAS: 7429-91-6 Summenformel: Dy Molekulargewicht (g/mol): 162.50 MDL-Nummer: MFCD00010982 InChI-Schlüssel: KBQHZAAAGSGFKK-UHFFFAOYSA-N PubChem CID: 23912 ChEBI: CHEBI:33377 IUPAC-Name: Dysprosium SMILES: [Dy]
Dysprosium-Folie, 0.025 mm (0.001 Zoll) dick, 0.999, Thermo Scientific Chemicals
CAS: 7429-91-6 Summenformel: Dy Molekulargewicht (g/mol): 162.50 MDL-Nummer: MFCD00010982 InChI-Schlüssel: KBQHZAAAGSGFKK-UHFFFAOYSA-N Synonym: ingot reo,disprosio,atom,ion,ingot,pieces,powder,foil, 3n,chips, 3n,ingot, 3n PubChem CID: 23912 ChEBI: CHEBI:33377 IUPAC-Name: Dysprosium SMILES: [Dy]
Dysprosium-Folie, 0.1 mm (0.004 Zoll) dick, 0.999, Thermo Scientific Chemicals
CAS: 7429-91-6 Summenformel: Dy Molekulargewicht (g/mol): 162.50 MDL-Nummer: MFCD00010982 InChI-Schlüssel: KBQHZAAAGSGFKK-UHFFFAOYSA-N Synonym: ingot reo,disprosio,atom,ion,ingot,pieces,powder,foil, 3n,chips, 3n,ingot, 3n PubChem CID: 23912 ChEBI: CHEBI:33377 IUPAC-Name: Dysprosium SMILES: [Dy]
Dysprosiumstab, 12.7 mm (0.5 in.) Durchmesser, 99.9 % (Metallbasis ohne Ta), Thermo Scientific Chemicals
CAS: 7429-91-6 Summenformel: Dy Molekulargewicht (g/mol): 162.50 MDL-Nummer: MFCD00010982 InChI-Schlüssel: KBQHZAAAGSGFKK-UHFFFAOYSA-N Synonym: ingot reo,disprosio,atom,ion,ingot,pieces,powder,foil, 3n,chips, 3n,ingot, 3n PubChem CID: 23912 ChEBI: CHEBI:33377 IUPAC-Name: Dysprosium SMILES: [Dy]
Dysprosiumstücke, 99.9 % (REO), Thermo Scientific Chemicals
CAS: 7429-91-6 Summenformel: Dy Molekulargewicht (g/mol): 162.50 MDL-Nummer: MFCD00010982 InChI-Schlüssel: KBQHZAAAGSGFKK-UHFFFAOYSA-N Synonym: ingot reo,disprosio,atom,ion,ingot,pieces,powder,foil, 3n,chips, 3n,ingot, 3n PubChem CID: 23912 ChEBI: CHEBI:33377 IUPAC-Name: Dysprosium SMILES: [Dy]
Dysprosiumfolie, 0.25 mm (0.01 Zoll) Dicke, 99.9 % (REO), Thermo Scientific Chemicals
CAS: 7429-91-6 Summenformel: Dy Molekulargewicht (g/mol): 162.50 MDL-Nummer: MFCD00010982 InChI-Schlüssel: KBQHZAAAGSGFKK-UHFFFAOYSA-N Synonym: ingot reo,disprosio,atom,ion,ingot,pieces,powder,foil, 3n,chips, 3n,ingot, 3n PubChem CID: 23912 ChEBI: CHEBI:33377 IUPAC-Name: Dysprosium SMILES: [Dy]
Dysprosium für AAS 1 g/L HNO₃ -Matrix, Fisher chemical™
CAS: 7429-91-6 Summenformel: Dy Molekulargewicht (g/mol): 162.50 MDL-Nummer: MFCD00010982 InChI-Schlüssel: KBQHZAAAGSGFKK-UHFFFAOYSA-N PubChem CID: 23912 ChEBI: CHEBI:33377 IUPAC-Name: Dysprosium SMILES: [Dy]
Dysprosiumpulver, -40 Mesh, 99.9 % (REO), Thermo Scientific Chemicals
CAS: 7429-91-6 Summenformel: Dy Molekulargewicht (g/mol): 162.50 MDL-Nummer: MFCD00010982 InChI-Schlüssel: KBQHZAAAGSGFKK-UHFFFAOYSA-N Synonym: ingot reo,disprosio,atom,ion,ingot,pieces,powder,foil, 3n,chips, 3n,ingot, 3n PubChem CID: 23912 ChEBI: CHEBI:33377 IUPAC-Name: Dysprosium SMILES: [Dy]
Dysprosium-Ingot, 99.8 % (REO), Thermo Scientific Chemicals
CAS: 7429-91-6 Summenformel: Dy Molekulargewicht (g/mol): 162.50 MDL-Nummer: MFCD00010982 InChI-Schlüssel: KBQHZAAAGSGFKK-UHFFFAOYSA-N Synonym: ingot reo,disprosio,atom,ion,ingot,pieces,powder,foil, 3n,chips, 3n,ingot, 3n PubChem CID: 23912 ChEBI: CHEBI:33377 IUPAC-Name: Dysprosium SMILES: [Dy]
Dysprosiumstücke, destilliert, dendritisch, 99.9 % (REO), Thermo Scientific Chemicals
CAS: 7429-91-6 Summenformel: Dy Molekulargewicht (g/mol): 162.50 MDL-Nummer: MFCD00010982 InChI-Schlüssel: KBQHZAAAGSGFKK-UHFFFAOYSA-N Synonym: ingot reo,disprosio,atom,ion,ingot,pieces,powder,foil, 3n,chips, 3n,ingot, 3n PubChem CID: 23912 ChEBI: CHEBI:33377 IUPAC-Name: Dysprosium SMILES: [Dy]
Dysprosiumstab, 6.35 mm (0.25 Zoll) Durchmesser, 99.7 % (Metallbasis ohne Ta), Thermo Scientific Chemicals
CAS: 7429-91-6 Summenformel: Dy Molekulargewicht (g/mol): 162.50 MDL-Nummer: MFCD00010982 InChI-Schlüssel: KBQHZAAAGSGFKK-UHFFFAOYSA-N Synonym: ingot reo,disprosio,atom,ion,ingot,pieces,powder,foil, 3n,chips, 3n,ingot, 3n PubChem CID: 23912 ChEBI: CHEBI:33377 IUPAC-Name: Dysprosium SMILES: [Dy]
Dysprosiumpulver, -200 Mesh, 99.9 % (REO), Thermo Scientific Chemicals
CAS: 7429-91-6 Summenformel: Dy Molekulargewicht (g/mol): 162.50 MDL-Nummer: MFCD00010982 InChI-Schlüssel: KBQHZAAAGSGFKK-UHFFFAOYSA-N Synonym: ingot reo,disprosio,atom,ion,ingot,pieces,powder,foil, 3n,chips, 3n,ingot, 3n PubChem CID: 23912 ChEBI: CHEBI:33377 IUPAC-Name: Dysprosium SMILES: [Dy]
Dysprosiumfolie, 0.62 mm (0.024 Zoll) Dicke, 99.8 % (REO), Thermo Scientific Chemicals
CAS: 7429-91-6 Summenformel: Dy Molekulargewicht (g/mol): 162.50 MDL-Nummer: MFCD00010982 InChI-Schlüssel: KBQHZAAAGSGFKK-UHFFFAOYSA-N Synonym: ingot reo,disprosio,atom,ion,ingot,pieces,powder,foil, 3n,chips, 3n,ingot, 3n PubChem CID: 23912 ChEBI: CHEBI:33377 IUPAC-Name: Dysprosium SMILES: [Dy]
- 1
- 2