CAS RN 627518-40-5
IUPAC Name:
—
Synonyme:
—
Molekulargewicht (g/mol):
—
Summenformel:
—
InChi Key:
—
SMILES:
—
1
–
1
von
1
Ergebnisse
1
R&D Systems™ JNJ 10198409
A Tocris product, part of R&D Systems. Potent PDGFRα and PDGFRβ inhbitor; also inhibits c-Abl, Lck, c-Src and Fyn kinases