CAS RN 2622-14-2
Trichyclohexylphosphin, technische Qualität, Thermo Scientific Chemicals
CAS: 2622-14-2 Summenformel: C18H33P Molekulargewicht (g/mol): 280.436 MDL-Nummer: MFCD00003853 InChI-Schlüssel: WLPUWLXVBWGYMZ-UHFFFAOYSA-N Synonym: tricyclohexylphosphine,tricyclohexyl phosphine,phosphine, tricyclohexyl,tricyclohexyl-phosphane,pcy3,tricyclohexylphosphine solution,tchp,p cy 3,phosphorus tricyclohexyl,tricyclohexylphosphine, dissolved in toluene concentration PubChem CID: 75806 IUPAC-Name: Tricyclohexylphosphan SMILES: C1CCC(CC1)P(C2CCCCC2)C3CCCCC3
Tricyclohexylphosphin, 1 M Lösung in Toluol, AcroSeal™, Thermo Scientific Chemicals
Tricyclohexylphosphin, 97 %, Thermo Scientific Chemicals ™
CAS: 2622-14-2 Summenformel: C18H33P Molekulargewicht (g/mol): 280.42 MDL-Nummer: MFCD00003853 InChI-Schlüssel: WLPUWLXVBWGYMZ-UHFFFAOYSA-N Synonym: tricyclohexylphosphine,tricyclohexyl phosphine,phosphine, tricyclohexyl,tricyclohexyl-phosphane,pcy3,tricyclohexylphosphine solution,tchp,p cy 3,phosphorus tricyclohexyl,tricyclohexylphosphine, dissolved in toluene concentration PubChem CID: 75806 IUPAC-Name: Tricyclohexylphosphan SMILES: C1CCC(CC1)P(C2CCCCC2)C3CCCCC3
Tricyclohexylphosphin, 96 %, Thermo Scientific Chemicals
CAS: 2622-14-2 Summenformel: C18H33P Molekulargewicht (g/mol): 280.436 MDL-Nummer: MFCD00003853 InChI-Schlüssel: WLPUWLXVBWGYMZ-UHFFFAOYSA-N Synonym: tricyclohexylphosphine,tricyclohexyl phosphine,phosphine, tricyclohexyl,tricyclohexyl-phosphane,pcy3,tricyclohexylphosphine solution,tchp,p cy 3,phosphorus tricyclohexyl,tricyclohexylphosphine, dissolved in toluene concentration PubChem CID: 75806 IUPAC-Name: Tricyclohexylphosphan SMILES: C1CCC(CC1)P(C2CCCCC2)C3CCCCC3
Tricyclohexylphosphin (90%), TRC
Hochreine organische Moleküle und analytische Standards, strategisch weltweit geliefert, um Innovation und wirtschaftlichen Erfolg zu ermöglichen.
Tricyclohexylphosphin, 98 %, Thermo Scientific Chemicals
CAS: 2622-14-2 Summenformel: C18H33P Molekulargewicht (g/mol): 280.436 MDL-Nummer: MFCD00003853 InChI-Schlüssel: WLPUWLXVBWGYMZ-UHFFFAOYSA-N Synonym: tricyclohexylphosphine,tricyclohexyl phosphine,phosphine, tricyclohexyl,tricyclohexyl-phosphane,pcy3,tricyclohexylphosphine solution,tchp,p cy 3,phosphorus tricyclohexyl,tricyclohexylphosphine, dissolved in toluene concentration PubChem CID: 75806 IUPAC-Name: Tricyclohexylphosphan SMILES: C1CCC(CC1)P(C2CCCCC2)C3CCCCC3