CAS RN 38869-47-5
IUPAC Name:
1-(4-methoxyphenyl)piperazine hydrochloridyl
Synonyme:
para-methoxyphenylpiperazine hydrochloridyl
Molekulargewicht (g/mol):
227.71
Summenformel:
C11H16ClN2O
InChi Key:
MFHKWSCQPZNUNF-UHFFFAOYSA-N
SMILES:
[Cl].COC1=CC=C(C=C1)N1CCNCC1
1
–
1
von
1
Ergebnisse
1
1-(4-Methoxyphenyl)-piperazin Dihydrochlorid, 97 %
CAS: 38869-47-5 Summenformel: C11H18Cl2N2O Molekulargewicht (g/mol): 265.178 MDL-Nummer: MFCD00012767 InChI-Schlüssel: MMXANKLJIHSIQH-UHFFFAOYSA-N Synonym: 1-4-methoxyphenyl piperazine dihydrochloride,1-4-methoxyphenyl piperazinedihydrochloride,piperazine, 1-4-methoxyphenyl-, dihydrochloride,1-4-methoxylphenyl-piperazine dihydrochloride,para-methoxyphenylpiperazine dihydrochloride,1-4-methoxyphenyl-piperazine dihydrochloride,4-methoxy-1-piperazinylbenzene, chloride, chloride,pubchem8579,acmc-1ct2c,ksc222g2n PubChem CID: 2723700 IUPAC-Name: 1-(4-Methoxyphenyl)Piperazin;Dihydrochlorid SMILES: COC1=CC=C(C=C1)N2CCNCC2.Cl.Cl