CAS RN 1983205-51-1
CAS RN 1983205-51-1
IUPAC Name:
potassium trifluoro[4-(pyrrolidine-1-carbonyl)phenyl]boranuide
Synonyme:
potassium trifluoro[4-(pyrrolidine-1-carbonyl)phenyl]boranuide
Molekulargewicht (g/mol):
281.13
Summenformel:
C11H12BF3KNO
InChi Key:
HEBRRMDWGHQMOE-UHFFFAOYSA-N
SMILES:
[K+].F[B-](F)(F)C1=CC=C(C=C1)C(=O)N1CCCC1
1
–
1
von
1
Ergebnisse
1
Kalium-4-(1-pyrrolidinylcarbonyl)-phenyltrifluorborat, 95 %, Thermo Scientific™
CAS: 1983205-51-1 Summenformel: C11H12BF3KNO Molekulargewicht (g/mol): 281.127 MDL-Nummer: MFCD16293904 InChI-Schlüssel: HEBRRMDWGHQMOE-UHFFFAOYSA-N Synonym: potassium 4-pyrrolidine-1-carbonyl phenyl trifluoroborate,potassium 4-1-pyrrolidinylcarbonyl phenyltrifluoroborate,potassium trifluoro 4-pyrrolidine-1-carbonyl phenyl boranuide,amtb215,potassium 4-pyrrolidine-1-carbonyl-phenyl trifluoroborate PubChem CID: 73995281 IUPAC-Name: Kalium;Trifluor-[4-(Pyrrolidin-1-Carbonyl)Phenyl]Boranuid SMILES: [B-](C1=CC=C(C=C1)C(=O)N2CCCC2)(F)(F)F.[K+]