CAS RN 13078-14-3
IUPAC Name:
1-(4-methylphenyl)piperazine hydrochloridyl
Synonyme:
1-(4-methylphenyl)piperazine hydrochloridyl
Molekulargewicht (g/mol):
211.71
Summenformel:
C11H16ClN2
InChi Key:
VCDXRVPYPWRHRI-UHFFFAOYSA-N
SMILES:
[Cl].CC1=CC=C(C=C1)N1CCNCC1
1
–
1
von
1
Ergebnisse
1
1-(p-Tolyl)piperazin-Dihydrochlorid, 97 %, Thermo Scientific™
CAS: 13078-14-3 Summenformel: C11H16ClN2 Molekulargewicht (g/mol): 211.71 MDL-Nummer: MFCD00035286 InChI-Schlüssel: VCDXRVPYPWRHRI-UHFFFAOYSA-N Synonym: 1-p-tolyl piperazine dihydrochloride,1-4-methylphenyl piperazine dihydrochloride,1-4-tolyl piperazine 2hcl,1-p-tolyl piperazine di hcl,4-methylphenyl piperazine, chloride, chloride,1-4-tolyl piperazine dihydrochloride,1-p-tolyl piperazine hcl,4-tolylpiperazine dihydrochloride,1-p-tolyl-piperazine dihydrochloride,1-4-tolyl-piperazine dihydrochloride PubChem CID: 83112 IUPAC-Name: 1-(4-methylphenyl)piperazine hydrochloridyl SMILES: [Cl].CC1=CC=C(C=C1)N1CCNCC1