Learn More
Agilent BioTek Spare Syringe Head/Plunger Assembly
Syringe Head/Plungers for use with the EL406, 406 FX, MultiFlo FX and MicroFill.
Marke: Agilent BioTek 7180046
Additional Details : Gewicht : 0.90718kg
Beschreibung
- Autoclavable
Spezifikation
Syringe Head | |
EL406 Microplate Washer, 406 FX Washer Dispenser |