Learn More
DWK Life Sciences Kimble™ Kontes™ Brand Valved 5mm O.D. NMR Tubes
Ideal for handling volatile, air-sensitive and toxic samples
Marke: DWK Life Sciences 897435-0000
Additional Details : Gewicht : 0.45000kg
Beschreibung
Ideal for handling volatile, air-sensitive and toxic samples
- Permit condensing of samples directly into NMR tube
- Greaseless, Hi-Vac™ valve lets you recover samples, then reseal; no need to permanently seal off, as with conventional tubes
- With glass tube and Viton™ O-ring for connection to a vacuum system
- Wall thickness is 0.01475 in. (0.37mm); Wall variation and camber depend, in part, on tube length
Spezifikation
0.013 mm | |
Borosilicate Glass | |
5 mm | |
0.0005 in. | |
Designed to improve sample handling during the investigation of volatile, air-sensitive, or toxic compounds | |
0.014 in. |
0.19 in. | |
177.8 mm | |
0.001 in. | |
>360MHz | |
7 in. | |
0.38 mm |