Learn More
Cytiva VAKUUM-VERBINDER 050/2/12
This Lower Part is a replacement part for Whatman™ GV 050 Series Vacuum Filtration Manifolds.
Marke: Cytiva 10442212
210.90 CHF gültig bis 2024-12-31
Benutzen Sie den Promotions-Code "23266" um Ihren Promotions-Preis zu erhalten.
Additional Details : Gewicht : 0.18000kg
Beschreibung
- Description: Replacement Part for GV 050 Series Glass Vacuum Filtration Manifolds
- For Use With (Application): Vacuum Filtration
Spezifikation
Autoclavable | |
Reusable | |
Borosilicate Glass | |
Vacuum Filtration | |
Je 1 | |
Filter Funnel |
Filter Funnel (Base Section, NS45) | |
Für GV050/2 | |
Non-sterile | |
GV050/2/12 | |
Gehäuse GV050 |