missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EIF3G Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | EIF3G |
---|---|
Konzentration | 0.23 mg/mL |
Anwendungen | Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16868053
|
Proteintech
11165-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen
|
|||||
16858053
|
Proteintech
11165-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen
|
|||||
Beschreibung
Component of the eukaryotic translation initiation factor 3 (eIF-3) complex, which is required for several steps in the initiation of protein synthesis. The eIF-3 complex associates with the 40S ribosome and facilitates the recruitment of eIF-1, eIF-1A, eIF-2:GTP:methionyl-tRNAi and eIF-5 to form the 43S preinitiation complex (43S PIC). The eIF-3 complex stimulates mRNA recruitment to the 43S PIC and scanning of the mRNA for AUG recognition. The eIF-3 complex is also required for disassembly and recycling of posttermination ribosomal complexes and subsequently prevents premature joining of the 40S and 60S ribosomal subunits prior to initiation. This subunit can bind 18S rRNA.Spezifikation
EIF3G | |
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O75821, Q5RK09, Q9Z1D1 | |
298700, 53356, 8666 | |
EIF3G Fusion Protein Ag1653 | |
Primary | |
-20°C | |
EIF3G |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
eIF 3 delta, eIF 3 RNA binding subunit, eIF3 delta, EIF3 P42, eIF3 p44, EIF3G, EIF3S4 | |
Eif3g | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |