missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEP164 Rabbit anti-Canine, Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | CEP164 |
---|---|
Konzentration | 0.29 mg/mL |
Anwendungen | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16851765
|
Proteintech
22227-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 18-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16841765
|
Proteintech
22227-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 04-06-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
CEP164 was initially identified as a centrosomal protein, but other studies have indicated that it also plays a role in the formation of primary cilia, the microtubule-based sensory antennae projecting from the surface of many eukaryotic cells as well as in DNA damage response acting as a mediator protein. CEP164 interacts with both ATR and ATM, proteins that trigger a number of cellular responses including the initiation of DNA damaged-induced cell cycle checkpoints. It is phosphorylated upon replication stress, ultraviolet (UV) radiation, and ionizing radiation silencing of CEP164 significantly reduces the DNA damage-induced phosphorylation of several proteins in the DNA damage-activated signaling cascade and compromises cell survival after UV damage. At least two isoforms of CEP164 are known to exist.Spezifikation
CEP164 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Canine, Human | |
Q9UPV0 | |
22897, 489389 | |
CEP164 Fusion Protein Ag17570 | |
Primary | |
-20°C | |
CEP164 |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
centrosomal protein 164kDa, Centrosomal protein of 164 kDa, CEP164, KIAA1052 | |
CEP164 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |